Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252226.1 | NP_001239155.1 | 8 | 1070 | |
NM_006622.3 | NP_006613.2 | 7 | 1112 | ||
GenBank mRNA | AF059617.1 | - | 7 | 1232 | |
AF223574.1 | - | 7 | 1055 | ||
AK027851.1 | - | 7 | 1056 | ||
AK098163.1 | - | 8 | 1038 | ||
AK297298.1 | - | 8 | 1015 | ||
AK312505.1 | - | 7 | 1060 | ||
BC013879.2 | - | 7 | 1045 | 3831747 | |
U85755.1 | - | 6 | 607 |