Gene Transcripts
- Gene Symbol
- MUM1
- Entrez Gene ID
- 84939
- Gene Name
- melanoma associated antigen (mutated) 1
- Gene Aliases
- EXPAND1, HSPC211, MUM-1
- Chromosome Location
- Chr. 19: 1354977 - 1378431 on Build GRCh38
- UniGene ID
- Hs.515016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032853.3 |
NP_116242.2
|
11, 12 |
1671 |
|
NR_024247.1 |
|
11, 12 |
1740 |
|
XM_011528380.2 |
XP_011526682.1
|
11, 12 |
1661 |
|
XM_011528382.2 |
XP_011526684.1
|
11, 12 |
1661 |
|
XM_011528383.1 |
XP_011526685.1
|
11, 12 |
1661 |
|
XM_011528384.2 |
XP_011526686.1
|
11, 12 |
1661 |
|
XM_011528385.2 |
XP_011526687.1
|
12, 13 |
1862 |
|
XM_017027401.1 |
XP_016882890.1
|
11, 12 |
1661 |
|
GenBank mRNA |
AK027774.1 |
- |
2, 3 |
140 |
|
AK075241.1 |
- |
5, 6 |
301 |
|
AK309092.1 |
- |
10, 11 |
1559 |
|
AK310721.1 |
- |
12, 13 |
1774 |
|
BC008098.1 |
- |
3, 4 |
155 |
|
BC082987.1 |
- |
2, 3 |
122 |
|
BC110874.1 |
- |
10, 11 |
1637 |
4933735 |
BC130443.1 |
- |
11, 12 |
1700 |
40146474 |
BC144138.1 |
- |
10, 11 |
1587 |
9052658 |
BC144147.1 |
- |
11, 12 |
1700 |
|
BQ071619.1 |
- |
7, 8 |
702 |
|
View Full Product Details