Gene Transcripts

Gene Symbol
TXNL4B
Entrez Gene ID
54957
Gene Name
thioredoxin like 4B
Gene Aliases
DLP, Dim2
Chromosome Location
Chr. 16: 72044261 - 72094316 on Build GRCh38
UniGene ID
Hs.134406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142317.1 NP_001135789.1 4 1882
NM_001142318.1 NP_001135790.1 4 1780
NM_001324355.1 NP_001311284.1 5 2031
NM_017853.2 NP_060323.1 4 1931
GenBank mRNA AK000518.1 - 4 1905
BC009646.1 - 4 1873 3458545

View Full Product Details