Gene Transcripts

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
Chromosome Location
Chr. 10: 113751262 - 113782433 on Build GRCh38
UniGene ID
Hs.228320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193434.1 NP_001180363.1 5, 6 789
NM_001193435.1 NP_001180364.1 3, 4 593
NM_024889.4 NP_079165.3 5, 6 689
NM_182601.1 NP_872407.1 4, 5 906
XM_005270162.2 XP_005270219.1 5, 6 828
XM_005270163.2 XP_005270220.1 5, 6 828
XM_011540171.1 XP_011538473.1 5, 6 828
XM_011540172.1 XP_011538474.1 6, 7 596
XM_011540173.1 XP_011538475.1 5, 6 828
XM_011540174.2 XP_011538476.1 5, 6 828
XM_011540175.1 XP_011538477.1 5, 6 767
XM_011540176.1 XP_011538478.1 3, 4 365
XM_017016666.1 XP_016872155.1 5, 6 828
GenBank mRNA AK126354.1 - 3, 4 365
AK128811.1 - 5, 6 689
AK292867.1 - 5, 6 561
BC037870.1 - 4, 5 911 5273287
FJ236306.1 - 4, 5 911

View Full Product Details