Gene Transcripts

Gene Symbol
PDZD8
Entrez Gene ID
118987
Gene Name
PDZ domain containing 8
Gene Aliases
PDZK8
Chromosome Location
Chr. 10: 117280489 - 117375467 on Build GRCh38
UniGene ID
Hs.501149
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173791.4 NP_776152.1 1 1094
XM_005269518.3 XP_005269575.1 1 1067
XM_011539265.1 XP_011537567.1 1 1067
XM_011539266.2 XP_011537568.1 1 1067
GenBank mRNA AK091746.1 - 1 1067
AY211929.1 - 1 163
BC028375.1 - 1 1056 4837939

View Full Product Details