Gene Transcripts
- Gene Symbol
- Rdx
- Entrez Gene ID
- 19684
- Gene Name
- radixin
- Gene Aliases
- AA516625
- Chromosome Location
- Chr. 9: 52047150 - 52088738 on Build GRCm38
- UniGene ID
- Mm.245746
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001104616.1 |
NP_001098086.1
|
3, 4 |
247 |
|
NM_001104617.1 |
NP_001098087.1
|
3, 4 |
653 |
|
NM_009041.3 |
NP_033067.2
|
3, 4 |
653 |
|
GenBank mRNA |
AK016711.1 |
- |
3, 4 |
324 |
|
AK088292.1 |
- |
3, 4 |
360 |
|
AK146423.1 |
- |
3, 4 |
311 |
|
AK156370.1 |
- |
3, 4 |
339 |
|
AK162204.1 |
- |
3, 4 |
225 |
|
AK168449.1 |
- |
3, 4 |
324 |
|
AK206703.1 |
- |
3, 4 |
191 |
|
AK217469.1 |
- |
3, 4 |
191 |
|
BC002041.1 |
- |
3, 4 |
298 |
|
BC026786.1 |
- |
3, 4 |
326 |
|
BC034438.1 |
- |
3, 4 |
479 |
|
BC053417.1 |
- |
3, 4 |
327 |
6707873 |
DV662898.1 |
- |
3, 4 |
247 |
|
X60672.1 |
- |
3, 4 |
266 |
|
View Full Product Details