Gene Transcripts

Gene Symbol
Selplg
Entrez Gene ID
20345
Gene Name
selectin, platelet (p-selectin) ligand
Gene Aliases
CD162, Psgl-1, Psgl1, Selp1, Selpl
Chromosome Location
Chr. 5: 113817798 - 113832638 on Build GRCm38
UniGene ID
Mm.332590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009151.3 NP_033177.3 2 307
GenBank mRNA AK089188.1 - 2 282
AK089214.1 - 2 233
AK155762.1 - 2 238
AK156933.1 - 2 238
AK158134.1 - 2 238
AK171693.1 - 2 251
AK172077.1 - 2 241
BC003874.1 - 2 239 3495181
CJ134610.1 - 2 308
X91144.1 - 1 156

View Full Product Details