Gene Transcripts

Gene Symbol
Psme3
Entrez Gene ID
19192
Gene Name
proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki)
Gene Aliases
AA410043, AU020960, Ki, PA28gamma, REGgamma, pa28g
Chromosome Location
Chr. 11: 101316251 - 101323531 on Build GRCm38
UniGene ID
Mm.288477
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011192.3 NP_035322.1 5 462
GenBank mRNA AK140597.1 - 5 483
AK150939.1 - 5 482
AK156079.1 - 5 432
AK171348.1 - 5 478
AK182764.1 - 3 206
AK183517.1 - 3 206
AK183726.1 - 3 167
AK184446.1 - 3 206
AK196904.1 - 3 206
AK198462.1 - 3 206
AK210357.1 - 3 202
BC015255.1 - 5 462 4221225
BC139818.1 - 5 410 40141268
CT010198.1 - 5 266
D87911.1 - 5 385
U60330.1 - 5 266

View Full Product Details