Gene Transcripts

Gene Symbol
Stra13
Entrez Gene ID
20892
Gene Name
stimulated by retinoic acid 13
Gene Aliases
Chromosome Location
Chr. 11: 120710941 - 120713903 on Build GRCm38
UniGene ID
Mm.259716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016665.2 NP_057874.2 5 472
GenBank mRNA AK076181.1 - 4 531
AK080500.1 - 5 444
AK170126.1 - 5 444
BC052335.1 - 6 722
CA320983.1 - 4 411
CJ236047.1 - 4 384
U95003.1 - 6 536
U95005.1 - 6 710

View Full Product Details