Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166482.1 | NP_001159954.1 | 7 | 675 | |
NM_172761.3 | NP_766349.1 | 7 | 769 | ||
GenBank mRNA | AK083317.1 | - | 7 | 709 | |
AK141278.1 | - | 7 | 604 | ||
AK188342.1 | - | 2 | 76 | ||
BC023782.1 | - | 7 | 746 | ||
BC026652.1 | - | 6 | 645 | ||
BC052066.1 | - | 6 | 2052 | 5708660 |