Gene Transcripts

Gene Symbol
Slc16a12
Entrez Gene ID
240638
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 12
Gene Aliases
3230401A21, AW210596
Chromosome Location
Chr. 19: 34668401 - 34748269 on Build GRCm38
UniGene ID
Mm.74636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172838.3 NP_766426.1 5 825
GenBank mRNA AK028284.1 - 5 823
AK029076.1 - 5 825
BC111902.1 - 4 778 40062303
BC113803.1 - 4 778 40062304

View Full Product Details