Gene Transcripts

Gene Symbol
Rab6b
Entrez Gene ID
270192
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
9630015C03, AI844641, C330006L04Rik, D9Bwg0185e
Chromosome Location
Chr. 9: 103112074 - 103185276 on Build GRCm38
UniGene ID
Mm.490252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173781.4 NP_776142.1 8 881
GenBank mRNA AK035893.1 - 8 840
AK082732.1 - 8 1006
BC059903.1 - 4 464
BC060618.1 - 8 881 6406134

View Full Product Details