Gene Transcripts

Gene Symbol
Acsm5
Entrez Gene ID
272428
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
C730019D22, C730027J19Rik
Chromosome Location
Chr. 7: 119526131 - 119543360 on Build GRCm38
UniGene ID
Mm.185183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178758.3 NP_848873.1 14 1903
GenBank mRNA AK050132.1 - 15 2000
AK050219.1 - 14 1928
AK050288.1 - 14 1923
AK050404.1 - 14 1963
AK050458.1 - 14 1865
AK149591.1 - 14 1923
BC095985.1 - 14 1903 4971395

View Full Product Details