Gene Transcripts
- Gene Symbol
- Prol1
- Entrez Gene ID
- 17830
- Gene Name
- proline rich, lacrimal 1
- Gene Aliases
- Muc10
- Chromosome Location
- Chr. 5: 88317312 - 88328817 on Build GRCm38
- UniGene ID
- Mm.200411
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008644.2 |
NP_032670.2
|
3 |
704 |
|
GenBank mRNA |
U37531.1 |
- |
4 |
587 |
|
View Full Product Details