Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164171.1 | NP_001157643.1 | 39 | 6015 | |
NM_010856.4 | NP_034986.1 | 39 | 5951 | ||
GenBank mRNA | BC110700.1 | - | 39 | 5997 | |
BC138027.1 | - | 38 | 5890 | 8861046 | |
M12291.1 | - | 2 | 50 | ||
M38127.1 | - | 1 | 14 | ||
M74751.1 | - | 2119 | |||
M76601.1 | - | 39 | 5915 |