Gene Transcripts

Gene Symbol
Eng
Entrez Gene ID
13805
Gene Name
endoglin
Gene Aliases
AI528660, AI662476, CD105, Endo, S-endoglin
Chromosome Location
Chr. 2: 32646532 - 32682669 on Build GRCm38
UniGene ID
Mm.225297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146348.1 NP_001139820.1 7 1191
NM_001146350.1 NP_001139822.1 7 1194
NM_007932.2 NP_031958.2 7 1194
GenBank mRNA AK151302.1 - 7 1164
AY679531.1 - 7 1194
BC029080.1 - 7 1108 4168250
EU330531.1 - 2 42
S69407.1 - 8 1059
X77952.1 - 7 1194

View Full Product Details