Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144512.2 | NP_653095.1 | 11 | 1248 | |
GenBank mRNA | AK149557.1 | - | 11 | 1256 | |
BC021606.1 | - | 2 | 106 | ||
BC023117.1 | - | 11 | 1236 | 4240641 | |
BC029637.1 | - | 11 | 1248 | 4195373 |