Gene Transcripts

Gene Symbol
Tomm7
Entrez Gene ID
66169
Gene Name
translocase of outer mitochondrial membrane 7 homolog (yeast)
Gene Aliases
1110020J08Rik, AW047273, Tom7
Chromosome Location
Chr. 5: 23838944 - 23844145 on Build GRCm38
UniGene ID
Mm.291868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025394.3 NP_079670.1 3 358
GenBank mRNA AK003864.1 - 3 321
AK008581.1 - 3 275
BC055352.1 - 3 336 5011269
BE647192.1 - 2 160
BU555283.1 - 3 361
BY773505.1 - 3 351

View Full Product Details