Gene Transcripts

Gene Symbol
Lrg1
Entrez Gene ID
76905
Gene Name
leucine-rich alpha-2-glycoprotein 1
Gene Aliases
1300008B03Rik, 2310031E04Rik, Lrg, Lrhg
Chromosome Location
Chr. 17: 56119678 - 56121946 on Build GRCm38
UniGene ID
Mm.348025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029796.2 NP_084072.1 2 1329
GenBank mRNA AB055885.1 - 2 1289
AF403429.1 - 2 1272
AK004940.1 - 1 1277
AK009555.1 - 1 1282
BC030733.1 - 2 1329 5123909
BC092077.1 - 2 1274 4975749

View Full Product Details