Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172892.3 | NP_766480.1 | 10, 11 | 1779 | |
GenBank mRNA | AK036354.1 | - | 10, 11 | 1781 | |
AK038937.1 | - | 10, 11 | 1784 | ||
BC089161.1 | - | 10, 11 | 1768 | 30440173 |