Gene Transcripts

Gene Symbol
Sepn1
Entrez Gene ID
74777
Gene Name
selenoprotein N, 1
Gene Aliases
1110019I12Rik, AI414492
Chromosome Location
Chr. 4: 134537892 - 134552166 on Build GRCm38
UniGene ID
Mm.29773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029100.2 NP_083376.2 12 1942
GenBank mRNA AK003819.1 - 2 506
BC043665.1 - 4 629
BU057838.1 - 4 629

View Full Product Details