Gene Transcripts

Gene Symbol
Rapgef5
Entrez Gene ID
217944
Gene Name
Rap guanine nucleotide exchange factor (GEF) 5
Gene Aliases
4932413M22, C86120, D030051B22Rik, GFR, Mrgef, mKIAA0277, mmr-gef
Chromosome Location
Chr. 12: 117516479 - 117759737 on Build GRCm38
UniGene ID
Mm.227642
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175930.5 NP_787126.3 15 1784
GenBank mRNA AK029995.1 - 15 1785
AK030016.1 - 15 1738
AK043108.1 - 3 427
AK083591.1 - 5 724
AK122234.1 - 3 546
BC046627.1 - 3 505 6494175

View Full Product Details