Gene Transcripts

Gene Symbol
Sec16b
Entrez Gene ID
89867
Gene Name
SEC16 homolog B (S. cerevisiae)
Gene Aliases
Lztr2, Rgpr, Rgpr-p117
Chromosome Location
Chr. 1: 157506733 - 157568425 on Build GRCm38
UniGene ID
Mm.87114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159986.1 NP_001153458.1 3 452
NM_033354.3 NP_203505.3 5 699
GenBank mRNA AB063356.1 - 3 452
AK132144.1 - 3 476
AK173302.1 - 5 680
BC059194.1 - 4 517 4222915

View Full Product Details