Gene Transcripts

Gene Symbol
Padi6
Entrez Gene ID
242726
Gene Name
peptidyl arginine deiminase, type VI
Gene Aliases
Pad6, Padi5, ePAD
Chromosome Location
Chr. 4: 140727355 - 140742643 on Build GRCm38
UniGene ID
Mm.271661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153106.2 NP_694746.2 8 920
GenBank mRNA AF529423.1 - 8 896
AK139616.1 - 8 921
BC053724.1 - 8 918 30051727

View Full Product Details