Gene Transcripts
- Gene Symbol
- Mfn2
- Entrez Gene ID
- 170731
- Gene Name
- mitofusin 2
- Gene Aliases
- D630023P19Rik, Fzo
- Chromosome Location
- Chr. 4: 147873586 - 147904909 on Build GRCm38
- UniGene ID
- Mm.154312
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB048831.1 |
- |
9, 10 |
1160 |
|
AF384100.1 |
- |
12, 13 |
1439 |
|
View Full Product Details