Gene Transcripts

Gene Symbol
Creb3l2
Entrez Gene ID
208647
Gene Name
cAMP responsive element binding protein 3-like 2
Gene Aliases
BBF2H7, C530025K05Rik, SCIRR69
Chromosome Location
Chr. 6: 37327255 - 37442186 on Build GRCm38
UniGene ID
Mm.169929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178661.4 NP_848776.2 2 636
GenBank mRNA AK035695.1 - 2 626
AK036712.1 - 2 628
AK041050.1 - 2 609
AK041669.1 - 2 636
AK042418.1 - 2 630
BC043466.1 - 2 598 5052843
BC138470.1 - 2 482 8861490
BC138471.1 - 2 482 8861491

View Full Product Details