Gene Transcripts

Gene Symbol
Rrp12
Entrez Gene ID
107094
Gene Name
ribosomal RNA processing 12 homolog (S. cerevisiae)
Gene Aliases
AA408556, AA536972, mKIAA0690
Chromosome Location
Chr. 19: 41862851 - 41896153 on Build GRCm38
UniGene ID
Mm.276044
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199447.2 NP_955518.1 23 2708
GenBank mRNA AK122346.1 - 22 3822
AK146550.1 - 23 2716
BC062977.1 - 23 2708 6853905

View Full Product Details