Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271725.1 | NP_001258654.1 | 6 | 1232 | |
NM_001271726.1 | NP_001258655.1 | 6 | 1268 | ||
NM_001271727.1 | NP_001258656.1 | 6 | 1537 | ||
NM_001271728.1 | NP_001258657.1 | 6 | 1335 | ||
NM_030706.3 | NP_109631.2 | 6 | 1331 | ||
GenBank mRNA | AB043550.1 | - | 6 | 1315 | |
AF220017.1 | - | 6 | 1180 | ||
AK039741.1 | - | 6 | 1329 | ||
AK045410.1 | - | 6 | 1328 | ||
AK079415.1 | - | 6 | 1482 | ||
AK141419.1 | - | 6 | 1178 | ||
AK147275.1 | - | 6 | 1445 | ||
AK147395.1 | - | 6 | 1326 | ||
AK164350.1 | - | 5 | 1034 | ||
AK220343.1 | - | 6 | 1536 | ||
BC058668.1 | - | 5 | 2143 | ||
BC058961.1 | - | 6 | 1335 | 6826143 |