Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023651.4 | NP_076140.2 | 2 | 380 | |
GenBank mRNA | AF329877.2 | - | 2 | 289 | |
AJ304506.1 | - | 2 | 313 | ||
AK011355.1 | - | 2 | 350 | ||
AK029099.1 | - | 2 | 350 | ||
AK029336.1 | - | 2 | 353 | ||
AK031710.1 | - | 2 | 380 | ||
AK031975.1 | - | 2 | 336 | ||
AK153438.1 | - | 2 | 329 | ||
AK169148.1 | - | 2 | 405 | ||
AK182904.1 | - | 1 | 126 | ||
BC023683.1 | - | 2 | 392 | 5343438 |