Gene Transcripts

Gene Symbol
Ankrd24
Entrez Gene ID
70615
Gene Name
ankyrin repeat domain 24
Gene Aliases
4631433D01Rik, 5730519E19Rik, D10Bur2e
Chromosome Location
Chr. 10: 81627795 - 81647612 on Build GRCm38
UniGene ID
Mm.304382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027480.3 NP_081756.4 17 2218
GenBank mRNA AK019475.1 - 4 884
AK173316.1 - 15 3184
BC017633.1 - 4 889
BC048236.1 - 16 2123 6313999

View Full Product Details