Gene Transcripts

Gene Symbol
Rusc1
Entrez Gene ID
72296
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
2210403N08Rik, AA408288, NESCA
Chromosome Location
Chr. 3: 89083975 - 89093363 on Build GRCm38
UniGene ID
Mm.27687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083807.1 NP_001077276.1 10 2744
NM_001083808.1 NP_001077277.1 9 1284
NM_028188.2 NP_082464.2 11 3155
GenBank mRNA AK032334.1 - 9 1774
AK039664.1 - 11 3083
AK042689.1 - 10 2692
AK170012.1 - 9 1298
AK170210.1 - 9 1247
BC052081.1 - 8 1478
BC056360.1 - 10 2682 6816267
BC057034.1 - 9 1350 5687517

View Full Product Details