Gene Transcripts

Gene Symbol
SSR4
Entrez Gene ID
6748
Gene Name
signal sequence receptor subunit 4
Gene Aliases
CDG1Y, TRAPD
Chromosome Location
Chr. X: 153794175 - 153798512 on Build GRCh38
UniGene ID
Hs.409223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204526.1 NP_001191455.1 6 518
NM_001204527.1 NP_001191456.1 6 541
NM_006280.2 NP_006271.1 5 491
NR_037927.1 7 712
XM_011531186.1 XP_011529488.1 6 400
XM_011531187.1 XP_011529489.1 6 400
XM_017029756.1 XP_016885245.1 6 1289
XM_017029757.1 XP_016885246.1 6 1024
GenBank mRNA AK057117.1 - 4 2074
AK094395.1 - 5 2194
AK290493.1 - 5 397
BC003371.1 - 5 371 2900758
BE791319.1 - 5 413
BG165852.1 - 6 525
BP202705.1 - 6 480
BT007192.1 - 5 367
CD672615.1 - 6 583
DB238237.1 - 5 490
X90583.1 - 5 397
Z69043.1 - 5 416

View Full Product Details