Gene Transcripts

Gene Symbol
Tmco5
Entrez Gene ID
67356
Gene Name
transmembrane and coiled-coil domains 5
Gene Aliases
1700095F04Rik, Tmco5a
Chromosome Location
Chr. 2: 116878691 - 116892500 on Build GRCm38
UniGene ID
Mm.131623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026104.4 NP_080380.1 6 494
GenBank mRNA AK007078.1 - 6 478
AK007251.1 - 6 492
BC049568.1 - 6 458 6742928
BU936444.1 - 6 497

View Full Product Details