Gene Transcripts

Gene Symbol
Qpctl
Entrez Gene ID
67369
Gene Name
glutaminyl-peptide cyclotransferase-like
Gene Aliases
1810019P04Rik, BB101812, gQC
Chromosome Location
Chr. 7: 19140217 - 19149196 on Build GRCm38
UniGene ID
Mm.234876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026111.3 NP_080387.2 7 1652
GenBank mRNA AK035587.1 - 7 1652
AK045687.1 - 7 1656
AK136746.1 - 794
BC058181.1 - 7 1675 5325516

View Full Product Details