Gene Transcripts

Gene Symbol
Sh3kbp1
Entrez Gene ID
58194
Gene Name
SH3-domain kinase binding protein 1
Gene Aliases
1200007H22Rik, 1700125L08Rik, 5830464D22Rik, AI447724, IN85, Ruk, Seta
Chromosome Location
Chr. X: 159627029 - 159975920 on Build GRCm38
UniGene ID
Mm.286495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135727.2 NP_001129199.1 3 649
NM_021389.6 NP_067364.2 2 214
GenBank mRNA AK004636.1 - 2 214
AK018032.1 - 2 231
AK020782.1 - 2 191
AK030839.1 - 2 215
AK038540.1 - 3 381
AK049182.1 - 3 650
AK080930.1 - 3 514
AK172022.1 - 3 630
BC034646.1 - 2 218
BC052863.1 - 2 163

View Full Product Details