Gene Transcripts
- Gene Symbol
- Rab37
- Entrez Gene ID
- 58222
- Gene Name
- RAB37, member RAS oncogene family
- Gene Aliases
- B230331O03Rik, B230354I04Rik
- Chromosome Location
- Chr. 11: 115091203 - 115162773 on Build GRCm38
- UniGene ID
- Mm.143789
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163753.1 |
NP_001157225.1
|
8, 9 |
984 |
|
NM_021411.4 |
NP_067386.3
|
8, 9 |
596 |
|
GenBank mRNA |
AB232633.1 |
- |
8, 9 |
575 |
|
AF233582.1 |
- |
8, 9 |
575 |
|
AK039716.1 |
- |
8, 9 |
590 |
|
AK040216.1 |
- |
8, 9 |
596 |
|
AK045995.1 |
- |
8, 9 |
587 |
|
AK046260.1 |
- |
8, 9 |
985 |
|
AK050483.1 |
- |
8, 9 |
590 |
|
BC050757.1 |
- |
8, 9 |
596 |
|
BC128020.1 |
- |
8, 9 |
594 |
40136290 |
View Full Product Details