Gene Transcripts

Gene Symbol
Rab37
Entrez Gene ID
58222
Gene Name
RAB37, member RAS oncogene family
Gene Aliases
B230331O03Rik, B230354I04Rik
Chromosome Location
Chr. 11: 115091203 - 115162773 on Build GRCm38
UniGene ID
Mm.143789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163753.1 NP_001157225.1 6 822
NM_021411.4 NP_067386.3 6 434
GenBank mRNA AB232633.1 - 6 413
AF233582.1 - 6 413
AK039716.1 - 6 428
AK040216.1 - 6 434
AK045995.1 - 6 425
AK046260.1 - 6 823
AK050483.1 - 6 428
BC050757.1 - 6 434
BC128020.1 - 6 432 40136290

View Full Product Details