Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025679.3 | NP_079955.1 | 2 | 257 | |
GenBank mRNA | AK014113.1 | - | 2 | 258 | |
AK014247.1 | - | 2 | 264 | ||
AK017452.1 | - | 2 | 259 | ||
AK017694.1 | - | 2 | 253 | ||
AK019264.1 | - | 2 | 243 | ||
AK033553.1 | - | 2 | 261 | ||
AK038295.1 | - | 1 | 456 | ||
BC004090.1 | - | 2 | 262 | 3593790 |