Gene Transcripts

Gene Symbol
Mkln1
Entrez Gene ID
27418
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Gene Aliases
A130067F06Rik, AU015903, BB154892
Chromosome Location
Chr. 6: 31398828 - 31511795 on Build GRCm38
UniGene ID
Mm.238038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013791.2 NP_038819.1 6 563
GenBank mRNA AK037967.1 - 6 564
AK077541.1 - 6 559
AK136123.1 - 3 246
AK155457.1 - 6 544
AK164822.1 - 6 559
AK165239.1 - 6 563
BC013703.1 - 6 547 3668925
U72194.1 - 6 546

View Full Product Details