Gene Transcripts

Gene Symbol
Ppp2r1a
Entrez Gene ID
51792
Gene Name
protein phosphatase 2, regulatory subunit A, alpha
Gene Aliases
6330556D22Rik, PP2A, PR65
Chromosome Location
Chr. 17: 20945454 - 20965905 on Build GRCm38
UniGene ID
Mm.294138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016891.3 NP_058587.1 12 1448
GenBank mRNA AB021743.1 - 12 1396
AK054239.1 - 12 1408
AK078135.1 - 12 1427
AK088776.1 - 12 1420
AK180835.1 - 3 139
AK186399.1 - 3 139
BC006606.1 - 12 1448 3157293
BC052678.1 - 12 1444 30062580

View Full Product Details