Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019635.2 | NP_062609.2 | 8 | 1109 | |
GenBank mRNA | AF271361.1 | - | 8 | 855 | |
AK002914.1 | - | 3 | 327 | ||
AY058922.1 | - | 8 | 995 | ||
BC037440.1 | - | 6 | 865 | 3585312 | |
BC049123.1 | - | 8 | 983 | 6813956 | |
U28726.1 | - | 8 | 988 |