Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199045.1 | NP_001185974.1 | 8 | 1500 | |
NM_001199046.1 | NP_001185975.1 | 8 | 1394 | ||
NM_001199047.1 | NP_001185976.1 | 7 | 1407 | ||
NM_019756.3 | NP_062730.1 | 7 | 1301 | ||
GenBank mRNA | AB024329.1 | - | 6 | 1075 | |
AB031043.1 | - | 7 | 1106 | ||
AF081568.1 | - | 7 | 1141 | ||
AK016086.1 | - | 8 | 1195 | ||
AK030214.1 | - | 8 | 1500 | ||
AK169454.1 | - | 7 | 1302 | ||
AK170304.1 | - | 7 | 1266 | ||
AK171546.1 | - | 7 | 1268 | ||
BC026465.1 | - | 7 | 1284 | 4484099 | |
BC132325.1 | - | 7 | 1134 | 40130602 |