Gene Transcripts
- Gene Symbol
- U2AF1
- Entrez Gene ID
- 7307
- Gene Name
- U2 small nuclear RNA auxiliary factor 1
- Gene Aliases
- FP793, RN, RNU2AF1, U2AF35, U2AFBP
- Chromosome Location
- Chr. 21: 43092956 - 43108291 on Build GRCh38
- UniGene ID
- Hs.365116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025203.1 |
NP_001020374.1
|
4, 5 |
341 |
|
NM_001025204.1 |
NP_001020375.1
|
5, 6 |
408 |
|
NM_006758.2 |
NP_006749.1
|
4, 5 |
341 |
|
GenBank mRNA |
AB451244.1 |
- |
4, 5 |
257 |
|
AF370386.1 |
- |
3, 4 |
1064 |
|
AJ627978.1 |
- |
4, 5 |
287 |
|
AK307560.1 |
- |
4, 5 |
290 |
|
BC001177.1 |
- |
4, 5 |
285 |
3534272 |
BC001923.1 |
- |
4, 5 |
285 |
2823296 |
BC005915.1 |
- |
5, 6 |
354 |
4295037 |
CB110007.1 |
- |
4, 5 |
341 |
|
M96982.1 |
- |
4, 5 |
295 |
|
- Gene Symbol
- U2AF1L5
- Entrez Gene ID
- 102724594
- Gene Name
- U2 small nuclear RNA auxiliary factor 1 like 5
- Gene Aliases
- Chromosome Location
- Chr. 21: 6484623 - 6499284 on Build GRCh38
- UniGene ID
- Hs.365116
- Species
- Homo sapiens
View Full Product Details