Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011912.3 | NP_036042.1 | 3 | 851 | |
GenBank mRNA | AB221556.1 | - | 3 | 822 | |
AF028715.1 | - | 3 | 825 | ||
BC090635.1 | - | 3 | 848 | 30942578 | |
BC116389.1 | - | 3 | 843 | 40101824 | |
BC116390.1 | - | 3 | 843 | 40101825 | |
Y17792.1 | - | 3 | 851 |