Gene Transcripts

Gene Symbol
Prol1
Entrez Gene ID
65182
Gene Name
proline rich, lacrimal 1
Gene Aliases
Muc10, RSM-1
Chromosome Location
Chr. 14: 21338464 - 21353533 on Build Rnor_6.0
UniGene ID
Rn.10113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133513.1 NP_598197.1 3 506
GenBank mRNA U03407.1 - 3 506

View Full Product Details