Gene Transcripts
- Gene Symbol
- Mylk2
- Entrez Gene ID
- 117558
- Gene Name
- myosin light chain kinase 2
- Gene Aliases
- Klmc, Mlck
- Chromosome Location
- Chr. 3: 148386185 - 148397851 on Build Rnor_6.0
- UniGene ID
- Rn.81191
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057209.1 |
NP_476557.1
|
3 |
616 |
|
GenBank mRNA |
J03886.1 |
- |
3 |
616 |
|
View Full Product Details