Gene Transcripts

Gene Symbol
Tgfbr2
Entrez Gene ID
81810
Gene Name
transforming growth factor, beta receptor 2
Gene Aliases
TGF-beta 2, Tgfbr2T
Chromosome Location
Chr. 8: 124310288 - 124399345 on Build Rnor_6.0
UniGene ID
Rn.9954
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031132.3 NP_112394.3 3 532
XM_008766690.1 XP_008764912.1 4 1088
GenBank mRNA GU067683.1 - 3 281
L09653.1 - 3 339
S67770.1 - 3 532

View Full Product Details