Gene Transcripts
- Gene Symbol
- Cubn
- Entrez Gene ID
- 80848
- Gene Name
- cubilin (intrinsic factor-cobalamin receptor)
- Gene Aliases
- GP280, IFCR, MGA1
- Chromosome Location
- Chr. 17: 80584921 - 80807181 on Build Rnor_6.0
- UniGene ID
- Rn.3236
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053332.2 |
NP_445784.1
|
16 |
2046 |
|
GenBank mRNA |
AF022247.1 |
- |
16 |
2046 |
|
View Full Product Details