Gene Transcripts
- Gene Symbol
- Timm23
- Entrez Gene ID
- 54312
- Gene Name
- translocase of inner mitochondrial membrane 23 homolog (yeast)
- Gene Aliases
- Chromosome Location
- Chr. 16: 8324038 - 8350067 on Build Rnor_6.0
- UniGene ID
- Rn.154974
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100362432
- Entrez Gene ID
- 100362432
- Gene Name
- mitochondrial import inner membrane translocase subunit Tim23-like
- Gene Aliases
- Chromosome Location
- Chr. 3: 82970670 - 82971887 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details