Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012503.2 | NP_036635.1 | 4 | 303 | |
XM_006246579.1 | XP_006246641.1 | 4 | 415 | ||
XM_008767768.1 | XP_008765990.1 | 4 | 435 | ||
XM_008767769.1 | XP_008765991.1 | 4 | 379 | ||
GenBank mRNA | BC088154.1 | - | 4 | 303 | |
FQ209763.1 | - | 4 | 339 | ||
FQ218553.1 | - | 4 | 379 | ||
FQ218644.1 | - | 4 | 339 | ||
FQ218824.1 | - | 4 | 339 | ||
FQ219107.1 | - | 4 | 376 | ||
FQ219710.1 | - | 4 | 420 | ||
M21770.1 | - | 3 | 205 |